EMD Millipore Calbiochem™ JAK Inhibitor I

JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC50 = 15 nM), JAK2 (IC50 = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC50 = 1 nM).

Manufacturer: EMD Millipore 420099500UG

 View more versions of this product

Catalog No. 42-009-9500UG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Protect from Light
    Target Proteins murine JAK1
    Molecular Formula C18H16FN3O
    Formula Weight 309.3g/mol
    Solubility DMSO
    Description 98% by HPLC
    Format Off-white solid
    Quantity 500μg
    CAS 457081-03-7

    • Packaged Under Inert Gas
    • Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit