EMD Millipore Calbiochem™ JNK Inhibitor II

JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC50 = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).

Manufacturer: EMD Millipore 42011950MG

 View more versions of this product

Catalog No. 42-011-950MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Protect from Light
    Target Proteins JNK 1, JNK 2, JNK 3
    Molecular Formula C14H8N2O
    Formula Weight 220.2g/mol
    Solubility DMSO
    Description 98% by HPLC
    Format Yellowish orange solid
    Quantity 50mg
    CAS 129-56-6

    • Packaged Under Inert Gas
    • Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit