Learn More
MilliporeSigma™ Calbiochem™ L3MBTL3 MBT Domain Blocker, UNC1215
A cell-permeable pyrrolidinyl-piperidinyl-benzamide that effectively competes against H4K20Me2 peptide for L3MBTL3 binding by targeting the first two L3MBTL3 MBT domains ( K d = 120nM in binding studies using recombinant 3MBT fragment), while exhibiting much reduced potency against L3MBTL1-H4K20Me1, 53BP1-H4K20Me2, MBTD1-H4K20Me1, or L3MBTL4-H2AK36Me1 interaction (IC50 ≥2μM; [peptide]
Supplier: MilliporeSigma™ 5.00557.0001
Description
A cell-permeable pyrrolidinyl-piperidinyl-benzamide that effectively competes against H4K20Me2 peptide for L3MBTL3 binding by targeting the first two L3MBTL3 MBT domains ( K d = 120nM in binding studies using recombinant 3MBT fragment), while exhibiting much reduced potency against L3MBTL1-H4K20Me1, 53BP1-H4K20Me2, MBTD1-H4K20Me1, or L3MBTL4-H2AK36Me1 interaction (IC50 ≥2μM; [peptide] = 150nM). Shown to increase GFP-L3MBTL fusion nucleus mobility (10nM to 1μM) in HEK293 transfectants. X-ray structural analysis reveals two 3MBT are bridged together by two UNC1215 molecules in a reciprocal fashion.Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Specifications
Off-white | |
Kme binding ligand of L3MBTL3 | |
529.7g/mol | |
Hygroscopic, Protect from light | |
10 mg |
98% by HPLC | |
C32H43N5O2 | |
DMSO | |
Powder |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.