EMD Millipore™ Calbiochem™ Laminin Nonapeptide

Active in cell adhesion and cell-surface receptor binding and in stimulating cell migration

Manufacturer: EMD Millipore™ 052337031MG

 View more versions of this product

Catalog No. 05-233-7031MG

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Quantity 1mg
    Format White to off-white lyophilized solid
    Sequence H-Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg-OH
    Storage Requirements -20°C
    Product Type Laminin Nonapeptide
    Purity ≥90% by HPLC

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit