Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Cdk1/2 Inhibitor II, NU6102
The Cdk1/2 Inhibitor II, NU6102, also referenced under CAS 444722-95-6, controls the biological activity of Cdk1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
$182.00 - $341.00
Specifications
Color | White |
---|---|
Description | 95% by HPLC |
Target Proteins | Cdk1/cyclin B, Cdk2/cyclin A3 |
CAS | 444722-95-6 |
Molecular Formula | C18H22N6O3S |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
21-771-31MG
|
MilliporeSigma™
2177131MG |
1 mg |
Each for $182.00
|
|
|||||
21-771-35MG
|
MilliporeSigma™
2177135MG |
5 mg |
Each for $341.00
|
|
|||||
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
White | |
Cdk1/cyclin B, Cdk2/cyclin A3 | |
C18H22N6O3S | |
DMSO | |
Solid |
95% by HPLC | |
444722-95-6 | |
402.5g/mol | |
Protect from light |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title