Learn More
MilliporeSigma Calbiochem LSD1 Inhibitor III, CBB1007
The LSD1 Inhibitor III, CBB1007 controls the biological activity of LSD1. This small molecule/inhibitor is primarily used for Cell Structure applications.
Manufacturer: MilliporeSigma 48947810MG
Description
Packaged Under Inert GasSpecifications
Supplied as a trifluoroacetate salt. | |
White solid | |
10mg | |
DMSO |
99% by HPLC | |
Hygroscopic Protect from Light |
|
C27H34N8O4·3CF3CO2H |