Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem LSD1 Inhibitor
The LSD1 Inhibitor, also referenced under CAS 927019-63-4, controls the biological activity of LSD1. This small molecule/inhibitor is primarily used for Cell Signaling applications.
Manufacturer: MilliporeSigma 48947610MG
Description
Packaged Under Inert GasSpecifications
95% by HPLC | |
Protect from Light | |
927019-63-4 | |
474.34g/mol |
Pale yellow to yellow solid | |
10mg | |
C15H40Cl4N8 | |
H2O |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title