Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ MALT1 Inhibitor I, MI-2
A cell-permeable chloromethyl amide that inhibits MALT1 proteolytic activity (IC50
Supplier: MilliporeSigma™ 5.00491.0001
Description
A cell-permeable chloromethyl amide that inhibits MALT1 proteolytic activity (IC50 = 5.84μM against MALT1 paracaspase domain dimer-catalyzed Ac-LRSR-AMC hydrolysis) in an irreversible manner via covalent interaction, displaying little or no activity against caspase-3, -8, or -9. Due to irreversible binding and effective intracellular accumulation (HBL-1 intracellular [MI-2]t = 2 h = 5μM; Medium [MI-2]t = 0 = 0.2μM), MI-2 is at least 250-times more effective than Z-VRPR-FMK in blocking constitutive NF-κB activity and c-Rel nuclear translocation in HBL-1 cultures (equal or better blockage by 200nM MI-2 than by 50μM Z-VRPR-FMK). Shown to selectively inhibit MALT1-dependent ABC-DLBCL proliferation, but not the MALT1-independent growths of ABC- (HLY-1 and U2932) or GCB-DLBCL (OCI-Ly1 and OCI-Ly7) cells, both in cultures in vitro (GI50 in 48 h = 200, 400, 400, and 500 nM, respectively, against HBL-1, OCI-Ly3, OCI-Ly10, and TMD8) and in mice in vivo (65%,, 61%, and 0% suppression, respectively, of HBL-1, TMD8, and OCI-Ly1 tumor expansion 24 h after 14 daily 25mg/kg/day i.p. dosages) without any apparent toxicity to mice.Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Specifications
Beige | |
MALT1 | |
DMSO | |
Solid |
97% by HPLC | |
C19H17Cl3N4O3 | |
Protect from Light, Protect from Moisture | |
10 mg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title