EMD Millipore Calbiochem™ MEK Inhibitor I

The MEK Inhibitor I, also referenced under CAS 297744-42-4, controls the biological activity of MEK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Manufacturer: EMD Millipore 4449375MG

 View more versions of this product

Catalog No. 44-493-75MG

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Storage Requirements Protect from Light
    Target Proteins MEK
    Molecular Formula C21H18N4OS
    Formula Weight 374.5g/mol
    Solubility DMSO or MeOH
    Description 95% by HPLC (sum of two isomers)
    Format White solid
    Quantity 5mg
    CAS 297744-42-4

    • Packaged Under Inert Gas
    • Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit