Learn More
EMD Millipore Calbiochem™ MEK Inhibitor VII Available on GSA/VA Contract for Federal Government customers only.
The MEK1/2 Inhibitor, also referenced under CAS 305350-87-2, controls the biological activity of MEK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Manufacturer:  EMD Millipore 4449395MG
 View more versions of this product
Catalog No. 44-493-95MG
  Check Availability
Add to cart
Specifications
Description
  • Packaged Under Inert Gas
  • Cell Permeable
Description & Specifications
Specifications
97% by HPLC
Protect from Light
MEK 1, MEK 2
C16H12F3N3S
DMSO or EtOH
Off-white solid
5mg
305350-87-2
335.4g/mol