EMD Millipore Calbiochem™ MEK Inhibitor VII

The MEK1/2 Inhibitor, also referenced under CAS 305350-87-2, controls the biological activity of MEK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Manufacturer: EMD Millipore 4449391MG

 View more versions of this product

Catalog No. 44-493-91MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Protect from Light
    Target Proteins MEK 1, MEK 2
    Molecular Formula C16H12F3N3S
    Formula Weight 335.4g/mol
    Solubility DMSO or EtOH
    Description 97% by HPLC
    Format Off-white solid
    Quantity 1mg
    CAS 305350-87-2

    • Packaged Under Inert Gas
    • Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit