Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem Met30 Antagonist, SMER3
The Met30 Antagonist, SMER3 controls the biological activity of Met30. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Manufacturer: MilliporeSigma 47585825MG
Description
Packaged Under Inert GasSpecifications
97% by HPLC | |
Protect from Light | |
C11H4N4O2 | |
DMSO |
Yellow solid | |
25mg | |
224.2g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title