Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Microcystin-LR, Microcystis aeruginosa
Cyclic heptapeptide toxin isolated from the freshwater cyanobacteria Microcystis aeruginosa.
Supplier: MilliporeSigma™ 475815500UG
Description
Cyclic heptapeptide toxin isolated from the freshwater cyanobacteria Microcystis aeruginosa . Potent inhibitor of protein phosphatase 1 (PP1) and protein phosphatase 2A (PP2A). Unlike Okadaic Acid (Cat. No. 495604), microcystin-LR is effective on both PP1 (IC50 = 1.7 nM) and PP2A (IC50 = 40 pM). Reacts with phosphatase in a two-step mechanism involving rapid binding and inactivation of the catalytic subunit followed by slower covalent interactions. Has no effect on protein kinases, making it useful for reducing the effect of contaminating phosphatase activities in protein kinase assays. A 2.5mm (250μg/100μl) solution of Microcystin-LR, Microcystis aeruginosa (Cat. No. 475821) in DMSO is also available.Specifications
Pale Yellow | |
PP1 | |
C49H74N10O12 | |
DMSO or MeOH | |
500 μg |
95% by HPLC | |
101043-37-2 | |
995.2g/mol | |
Lyophilized |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction