Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ MIF Antagonist, ISO-1
The MIF Antagonist, ISO-1, also referenced under CAS 478336-92-4, controls the biological activity of MIF.
Supplier: MilliporeSigma™ 4758375MG
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Light Yellow | |
MIF tautomerize | |
C12H13NO4 | |
DMSO | |
Solid |
98% by HPLC | |
478336-92-4 | |
235.1g/mol | |
Protect from light | |
5 mg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction