Learn More
MilliporeSigma™ Calbiochem™ MIF Antagonist IV, RDR 03785
The MIF Antagonist IV, RDR 03785 controls the biological activity of MIF. This small molecule/inhibitor is primarily used for Cell Structure applications.
Manufacturer:  MilliporeSigma™ 47584710MG
Catalog No. 47-584-710MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
97% by HPLC
Protect from Light
C19H18F3NO4
DMSO
Cream solid
10mg
381.4g/mol