Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MIF Antagonist V, p425

The MIF Antagonist V, p425, also referenced under CAS 2610-05-1, controls the biological activity of MIF. This small molecule/inhibitor is primarily used for Cell Structure applications.

Manufacturer:  MilliporeSigma™ 4758482GM

Catalog No. 47-584-82GM


Add to cart

Description

Description

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach
Specifications

Specifications

95% by HPLC
Hygroscopic
Protect from Light
MIF
992.8g/mol
H2O
992.79
Blue solid
2gm
2610-05-1
C34H24N6Na4O16S4
UPKAWFACSJWKND-ZXFFUEEESA-J
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit