Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem Miltefosine
An ether lipid analog that acts as an inhibitor of CTP:phosphocholine cytidyltransferase and displays remarkable antiproliferative effect both in vitro and in vivo.
Manufacturer: MilliporeSigma 47584150MG
Description
Packaged Under Inert GasSpecifications
98% by TLC | |
Protect from Light | |
CTP:phosphocholine cytidyltransferase | |
C21H46NO4P | |
DMSO or EtOH |
Crystalline solid | |
50mg | |
58066-85-6 | |
407.6g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title