Learn More
MilliporeSigma Calbiochem Minocycline, Hydrochloride
Semi-synthetic tetracycline derivative effective against tetracycline-resistant staphylococci.
Manufacturer: MilliporeSigma 47584350MG
Description
Semi-synthetic tetracycline derivative effective against tetracycline-resistant staphylococci. A matrix metalloproteinase (MMP) inhibitor shown to inhibit tumor-induced angiogenesis. Inhibits stromelysin (MMP-3; IC50 = 290µM) but does not inhibit other MMPs such as fibroblast collagenase or gelatinase. Potently inhibits PARP-1 (K i = 13.4nM) in a NAD + - competitive manner. Exhibits anti-inflammatory effects and suppresses T cell proliferation.Specifications
98% by TLC | |
50mg | |
13614-98-7 | |
493.9g/mol |
Dark yellow solid | |
Metalloproteinase (MMP) | |
C23H27N3O7·HCl | |
H2O |