Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mitochondrial Fusion Promoter, M1

A cell-permeable phenylhydrazone that restores mitochondrial tubular network formation in MEF lacking either of the two outer mitochondrial membrane.

Manufacturer:  MilliporeSigma™ 47585925MG

Catalog No. 47-585-925MG


Add to cart

Description

Description

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
Specifications

Specifications

98% by HPLC
Protect from Light
219315-22-7
DMSO
Off-white solid
25mg
364.1g/mol
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit