Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem (+)-MK 801 Maleate
A highly potent, selective, and non-competitive NMDA receptor antagonist that acts by binding to a site located within the NMDA-associated ion channel, thus preventing Ca2+ flux.
Manufacturer: MilliporeSigma 47587810MG
Description
A highly potent, selective, and non-competitive NMDA receptor antagonist that acts by binding to a site located within the NMDA-associated ion channel, thus preventing Ca2+ flux. An effective anti-ischemic agent in several animal models. Has been used to develop an in vivo model of NMDA receptor-dependent apoptotic neurodegenerative disease in developing rat brains. Has also been used to develop an in vitro model of NMDA receptor-specific Ca2+ entry induced seizures.Specifications
98% by HPLC | |
Protect from Moisture | |
NMDA receptor | |
C16H15N·C4H4O4 | |
EtOH or H2O |
White solid | |
10mg | |
77086-22-7 | |
337.4g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title