Learn More
MilliporeSigma™ Calbiochem™ MNK1/2 Inhibitor II, ETP-45835
The MNK1/2 Inhibitor II, ETP-45835 controls the biological activity of MNK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Manufacturer:  MilliporeSigma™ 45486210MG
Catalog No. 45-486-210MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
98% by HPLC
Hygroscopic
Protect from Light
C13H16N4·2HCl
H2O
White solid
10mg
301.2g/mol