Learn More
MilliporeSigma™ Calbiochem™ mutant p53 Reactivator, RETRA
The mutant p53 Reactivator, RETRA, also referenced under CAS 1036069-26-7, controls the biological activity of mutant p53. This small molecule/inhibitor is primarily used for Activators/Inducers applications.
Manufacturer:  MilliporeSigma™ 5061645MG
Catalog No. 50-616-45MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
98% by HPLC
Hygroscopic
Protect from Light
1036069-26-7
305.8g/mol
Off-white solid
5mg
C11H11NO3S2·HCl
DMSO