Learn More
MilliporeSigma™ Calbiochem™ N-Acetyl-α-D-Glucosamine
Acceptor substrate for galactosyltransferases
Manufacturer:  MilliporeSigma™ 107925GM
Catalog No. 10-792-5GM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Inhibits lectin WGA and β-N-acetylhexosaminidase
  • When added to medium, it improves recovery of Haemophilus influenza from sputum
Specifications
Specifications
N-Acetyl-α-D-Glucosamine
25 g
Solid
White to Off-white