Learn More
MilliporeSigma™ Calbiochem™ NAD+, Free Acid
Acts as major electron acceptor molecule in biological oxidations
Supplier: MilliporeSigma™ 48191110GM
Specifications
-20°C | |
NAD+, Free Acid | |
10 g |