Learn More
MilliporeSigma™ Calbiochem™ NADH, Disodium Salt
Click to view available options
Quantity:
500 mg
Description
Unstable in solution; reconstitute just prior to use.
Specifications
Specifications
Content And Storage | -20°C |
Additive | NADH, Disodium Salt |
Quantity | 500 mg |