Learn More
MilliporeSigma™ Calbiochem™ NBI-31772
A non-peptide ligand that exhibits high-affinity (Ki »1 to 17 nM) towards all six human insulin-like growth factor-I binding proteins (IGFBPs).
Manufacturer:  MilliporeSigma™ 4798305MG
Catalog No. 47-983-05MG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
95% by HPLC
Protect from Light
Human insulin-like growth factor-I binding proteins (IGFBPs).
C17H11NO7
DMSO
Greenish-yellow solid
5mg
374620-70-9
341.3g/mol