Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem NEMO-Binding Domain Binding Peptide, Cell-Permeable
A cell-permeable Antennapedia-NBD (wild type) fusion peptide that exhibits anti-inflammatory properties.
Manufacturer: MilliporeSigma 480025500UG
Description
- Packaged Under Inert Gas
- Peptide Sequence: H-Asp-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Thr-Ala-Leu-Asp-Trp-Ser-Trp-Leu-Gln-Thr-Glu-OH
- Cell Permeable
Specifications
Supplied as a trifluoroacetate salt. | |
White lyophilized solid | |
500μg | |
C170H259N49O42S | |
DMSO |
95% by HPLC | |
Hygroscopic | |
cytokine-induced NF-κB activation and NF-κB-dependent gene expression | |
3693.3g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title