Learn More
MilliporeSigma™ Calbiochem™ Neurodazine
A cell-permeable trisubstituted imidazole compound that induces neurogenesis of non-pluripotent C2C12 myoblasts as well as cells derived from mature human skeletal muscle cultures (2µM for 7-10 days).
Manufacturer:  MilliporeSigma™ 48070310MG
Catalog No. 48-070-310MG
Add to cart
Description
Description
A cell-permeable trisubstituted imidazole compound that induces neurogenesis of non-pluripotent C2C12 myoblasts as well as cells derived from mature human skeletal muscle cultures (2µM for 7-10 days). Shown to preferentially upregulate genes involved in neurogenesis, but not those pertaining to myogenesis, in C2C12 myoblasts.
Specifications
Specifications
98% by HPLC
Protect from Light
1135318-08-9
456.9g/mol
Yellow film
10mg
C27H21ClN2O3
DMSO or EtOH