Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem Neuronal Differentiation Inducer, Isobavachin
A cell-permeable, 8-prenyl flavone that is shown to induce differentiation of mouse embryonic stem (ES) cells into neuronal cells and astrocytes, at 100 nM.
Manufacturer: MilliporeSigma 4807485MG
Description
Packaged Under Inert GasSpecifications
95% by HPLC | |
Protect from Light | |
C20H20O4 | |
DMSO |
Pale yellow solid | |
5mg | |
324.37g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title