Learn More
MilliporeSigma™ Calbiochem™ Neuronal Differentiation Inducer IV
A cell-permeable quinoxaline compound that is shown to induce neuronal differentiation in PC12 and rat MSC (mesenchymal stem cell) cultures with high efficiency.
Manufacturer:  MilliporeSigma™ 4807465MG
Catalog No. 48-074-65MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
95% by HPLC
Protect from Light
C18H17N3O3
DMSO
Off-white solid
5mg
323.4g/mol