Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Akt Inhibitor IV
The Akt Inhibitor IV, also referenced under CAS 681281-88-9, controls the biological activity of Akt.
$182.00 - $654.00
Specifications
Color | Yellow |
---|---|
Description | 98% by HPLC |
Target Proteins | Akt |
CAS | 681281-88-9 |
Molecular Formula | C31H27IN4S |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
12-401-11MG
|
MilliporeSigma™
1240111MG |
1 mg |
Each for $182.00
|
|
|||||
12-401-15MG
|
MilliporeSigma™
1240115MG |
5 mg |
Each for $654.00
|
|
|||||
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Yellow | |
Akt | |
C31H27IN4S | |
DMSO | |
Solid |
98% by HPLC | |
681281-88-9 | |
614.6g/mol | |
Hygroscopic, Protect from light |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title