Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Neurotrophin Antagonist, Y1036

Catalog No. 48087010MG
Click to view available options
Quantity:
10 mg

The Neurotrophin Antagonist, Y1036 controls the biological activity of Neurotrophin. This small molecule/inhibitor is primarily used for Neuroscience applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 90% by HPLC
Molecular Formula C17H14N2O6S3
Formula Weight 438.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.