Learn More
MilliporeSigma™ Calbiochem™ NF-κB Activation Inhibitor X, Methysticin
Methysticin has been identified from kava to have potent NF-κB inhibitory activity and minimum toxicity
Manufacturer:  MilliporeSigma™ 4804585MG
Catalog No. 48-045-85MG
Add to cart
Description
Description
Methysticin has been identified from kava to have potent NF-κB inhibitory activity and minimum toxicity. It is shown to inhibit NF-κB (IC50 = 0.19µg/ml) 100 times more potently than traditional kava in A549 cells in an in vitro luciferase-based assay. It is also reported to suppress the elevation of IKKα, p65, and COX-2 and the degradation of IκBα in A549 cultures stimulated with TNF-alpha; through Western Blot analysis.
Specifications
Specifications
99% by HPLC
5mg
274.3g/mol
White solid
C15H14O5
DMSO