Learn More
MilliporeSigma™ Calbiochem™ NF449
A potent Gsα-subunit-selective G-protein antagonist. Suppresses GTPγS binding to Gsα-s-subunit (IC50 = 140 nM). A highly potent P2X1 receptor antagonist (IC50 = 280 pM).
Manufacturer:  MilliporeSigma™ 48042010MG
Catalog No. 48-042-010MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
95% by HPLC
Hygroscopic
Protect from Light
Gsα-subunit-selective G-protein antagonist
C41H24N6O29S8·8Na
dH2O
White solid
10mg
389142-38-5
1505.1g/mol