Learn More
MilliporeSigma™ Calbiochem™ Niclosamide
A salicylanilide anthelmintic drug that acts as a potent and selective Stat3- and Wnt-signaling pathway inhibitor.
Manufacturer:  MilliporeSigma™ 4819091GM
Catalog No. 48-190-91GM
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
97% by HPLC
Protect from Light
50-65-7
327.1g/mol
Pale yellow solid
1gm
C13H8Cl2N2O4
DMSO or EtOH