Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Nitric Oxide Assay Kit (Colorimetric)
Used for quantitative measurement of total nitric oxide by nitrate reductase.
Supplier: MilliporeSigma™ 4826501KTT
Includes: Recombinant Zea mays Nitrate Reductase, NADH, Nitrate Standard, Color Reagents, Buffer, 96-Well Plate, and user protocol
Description
EMD Millipore Calbiochem™ Nitric Oxide Assay Kit (Colorimetric) is based on enzymatic conversion of nitrate to nitrite.
- Colorimetric detection Spectrophotometric quantitation of nitrite levels using Griess Reagent
- Format: 96 well plate
- TotalAssayTime: 1 to 1.5 hr
- Sufficient for 80 Tests
- Sample Type: Aqueous solutions
Specifications
Colorimetric | |
1 Kit | |
Spectrophotometric measurement of the nitric oxide | |
2° to 8°C |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction