Learn More
MilliporeSigma™ Calbiochem™ NMDA Receptor NR2BCT(1292-1304)-TAT
A 13-mer NMDA receptor 2B (NR2B) subunit carboxy terminal (1292-1304) derived peptide that specifically binds DAPK1 in a reversible manner and antagonizes the activated DAPK1-NR2B interaction and the potentiation of NMDAR function
Manufacturer:  MilliporeSigma™ 4545852MG
Catalog No. 45-458-52MG
Add to cart
Description
Description
A 13-mer NMDA receptor 2B (NR2B) subunit carboxy terminal (1292-1304) derived peptide that specifically binds DAPK1 (death-associated protein kinase 1) in a reversible manner and antagonizes the activated DAPK1-NR2B interaction and the potentiation of NMDAR function with no effect on either the catalytic activity of DAPK1 or the NMDA receptor physiology. Blocks Ca2+ influx through extrasynaptic NMDA receptors (at 50µM) and the blood-brain-barrier permeable version, namely NR2BCT(1292-1304) attached to TAT(47-57) via a spacer (6-aminohexanoic acid) protects cortical neurons from NMDA receptor-mediated insults and reduces brain infarction and improve neurological functions in mice (10mg/kg, i.v.).
Specifications
Specifications
Supplied as a trifluoroacetate salt
White powder
2mg
3382g/mol
98% by HPLC
Hygroscopic
Protect from Light
C143H254N64O32
H2O