Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem Nodinitib-1
A cell-permeable aminobenzimidazole compound that acts as a potent, specific and reversible inhibitor of NOD1-induced NF-κB activation.
Manufacturer: MilliporeSigma 48800410MG
Description
Packaged Under Inert GasSpecifications
99% by HPLC | |
Protect from Light | |
C14H13N3O2S | |
DMSO |
White powder | |
10mg | |
287.3g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title