Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem Nogo-66 (1-40) Antagonist Peptide, Synthetic
The Nogo-66 (1-40) Antagonist Peptide, Synthetic controls the biological activity of Nogo-66 (1-40). This small molecule/inhibitor is primarily used for Neuroscience applications.
Manufacturer: MilliporeSigma 4882301MG
Description
- Packaged Under Inert Gas
- Peptide Sequence: Ac-Arg-Ile-Tyr-Lys-Gly-Val-Ile-Gln-Ala-Ile-Gln-Lys-Ser-Asp-Glu-Gly-His-Pro-Phe-Arg-Ala-Tyr-Leu-Glu-Ser-Glu-Val-Ala-Ile-Ser-Glu-Glu-Leu-Val-Gln-Lys-Tyr-Ser-Asn-Ser-NH2
Specifications
Supplied as a trifluoroacetate salt. | |
White solid | |
1mg | |
C206H324N56O65 | |
DMSO |
95% by HPLC | |
Hygroscopic Protect from Light |
|
Nogo-66 receptor (NgR) | |
4625.2g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title