Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem NOX Inhibitor III, VAS2870
The NOX Inhibitor III, VAS2870, also referenced under CAS 722456-31-7, controls the biological activity of NOX.
Manufacturer: MilliporeSigma 49200010MG
Description
Packaged Under Inert GasSpecifications
95% by HPLC | |
Protect from Light | |
722456-31-7 | |
360.4g/mol |
Tan solid | |
10mg | |
C18H12N6OS | |
DMSO |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title