Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem Nrf2 Activator
The Nrf2 Activator modulates the biological activity of Nrf2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Manufacturer: MilliporeSigma 49204025MG
Description
Packaged Under Inert GasSpecifications
98% by HPLC | |
Protect from Light | |
C17H13F3O2 | |
DMSO or EtOH |
Yellow syrup | |
25mg | |
306.3g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title