Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem Nur77 LBD Antagonist, TMPA
A cell-permeable phenylacetate compound that blocks Nur77 (TR3) LKB1 nuclear-sequestering function by antagonizing against Nur77-LKB1 interaction via direct binding at the LKB1 LBD/ligand binding domain ( K d
Manufacturer: MilliporeSigma 49291010MG
Description
A cell-permeable phenylacetate compound that blocks Nur77 (TR3) LKB1 nuclear-sequestering function by antagonizing against Nur77-LKB1 interaction via direct binding at the LKB1 LBD/ligand binding domain ( K d = 0.14 against 5µM LKB1 binding to 5µM full-length Nur77). Shown to induce LKB1 Ser428 phosphorylation (10 and 20µM for 6 h) and LKB1 nuclear-to-cytosol translocation in hepatic LO2 cells, resulting in enhanced phosphorylations of LKB1 cytosolic effectors AMPKα and MARK1/2/3 without affecting Nur77 transactivation activity. Daily i.p. dosing (50mg/kg) is shown to increase liver LKB1 Ser428/AMPKα Thr172 phosphorylations in diabetic db/db mice in vivo , effectively improving animal glucose tolerance. Also reported to lower fasting blood glucose level in STZ- (Cat. No. 572201) treated, high fat diet-fed type II diabetic mice.Specifications
97% by HPLC | |
Protect from Light | |
Nur77 | |
380.5g/mol |
Off-white solid | |
10mg | |
C21H32O6 | |
DMSO |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title