Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem Olomoucine II
A cell-permeable 2,6,9-trisubstituted purine analog that acts as a potent, reversible, and ATP-competitive inhibitor of Cdk1/B.
Manufacturer: MilliporeSigma 4956215MG
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
95% by HPLC | |
Protect from Light | |
Cdk1/B | |
C19H26N6O2 | |
DMSO or EtOH |
Colorless solid | |
5mg | |
500735-47-7 | |
370.5g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title