Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem Omi/HtrA2 Protease Inhibitor, Ucf-101
The Omi/HtrA2 Protease Inhibitor, Ucf-101, also referenced under CAS 313649-08-0, controls the biological activity of Omi/HtrA2 Protease. This small molecule/inhibitor is primarily used for Cancer applications.
Manufacturer: MilliporeSigma 49615010MG
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
97% by HPLC | |
Protect from Light | |
pro-apoptotic, heat-inducible, mitochondrial serine protease Omi/HtrA2 | |
C27H17N3O5S | |
DMSO |
Orange solid | |
10mg | |
313649-08-0 | |
495.5g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title