EMD Millipore™ Calbiochem™ OmniPur™ Acrylamide: Bis-acrylamide 19:1, 40% Solution

Suitable for DNA and RNA separation and sequencing

Manufacturer: EMD Millipore™ 13101L

 View more versions of this product

Catalog No. 13-101-L

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Product Type Acrylamide: Bis-acrylamide
    Quantity 1L
    Storage Requirements 15-30°C
    Format Clear, colorless solution

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit