EMD Millipore™ Calbiochem™ OmniPur™ Acrylamide: Bis-acrylamide 37.5:1, 40% Solution
Suitable for protein seperations
Manufacturer: EMD Millipore™ 1500500ML
 View more versions of this product
Catalog No. 15-005-00ML
  Check Availability
Add to Cart
Disclaimers:
For Research Use Only
    Specifications
    Product Type Acrylamide: Bis-acrylamide
    Quantity 500mL
    Format Clear, colorless solution
  • Description & Specifications
Related Products
Product Content Feedback