Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ OmniPur™ Bradford Method Protein Assay Kit
Performs protein assay.
Supplier: MilliporeSigma™ 27401KIT
Description
EMD Millipore Calbiochem™ OmniPur™ Bradford Method Protein Assay Kit contains Bradford Reagent, Sodium Chloride BSA Solution.
- Contains: Bradford Reagent, 0.15M Sodium Chloride, 0.5mg/mL BSA Solution
- Specifications:
- BSA Solution BSA Content: 0.45-0.55mg/mL
- BSA Solution Protease: None Detected
- Bradford Reagent Pattern: Passes test
- Bradford Reagent Protease: None Detected
- Sodium Chloride Solution Concentration: 130-170mMol/l
Specifications
2° to 8°C | |
Bradford Method Protein Assay Kit | |
1 Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title