Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem OmniPur BSA, Molecular Biology Grade, Fraction V, Cold Alcohol Isolation
Used for multiple biological applications.
Manufacturer: MilliporeSigma 29055GM
Description
- Molecular Biology Grade
- Specifications:
- Ash: ≤0.5%
- Assay (by electrophoresis): ≥99%
- DNase: None Detected
- Heavy metals (as Pb): ≤0.005%
- Loss on drying: ≤5%
- pH (7%, water) at 25°C: 5.0 to 5.4
- Protease: None Detected
- RNase: None Detected
- CAS: 9048-46-8
- Molecular Biology Grade
- Color: White
Specifications
BSA | |
5 g |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title