Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ OmniPur™ Ethidium Bromide
Suitable for fluorescent staining of nucleic acids.
Supplier: MilliporeSigma™ 43101GM
Description
EMD Millipore Calbiochem™ OmniPur™ Ethidium Bromide facilitates purification of DNA in Cesium Chloride gradient.
- Fluorescent stain
- Molecular biology grade
- Specifications:
- Assay (dry basis): ≥98.0%
- DNase: None Detected
- Loss on drying: ≤8.0%
- Protease: None Detected
- RNase: None Detected
- CAS: 1239-45-8
- Molecular formula: C21H20N3Br
Specifications
Dark Red/Purple | |
Fluorescent DNA/RNA Stain | |
1 g |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction