Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Ethidium Bromide

Suitable for fluorescent staining of nucleic acids.

Specifications

Color Dark Red/Purple
Description Ethidium Bromide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
43-405-GM
View Documents
MilliporeSigma™
43405GM
5 g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
43-101-GM
View Documents
MilliporeSigma™
43101GM
1 g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

EMD Millipore Calbiochem™ OmniPur™ Ethidium Bromide facilitates purification of DNA in Cesium Chloride gradient.

  • Fluorescent stain
  • Molecular biology grade
  • Specifications:
  • Assay (dry basis): ≥98.0%
  • DNase: None Detected
  • Loss on drying: ≤8.0%
  • Protease: None Detected
  • RNase: None Detected
Specifications

Specifications

Dark Red/Purple
Ethidium Bromide
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit